Få mere at vide
Thermo Scientific™ Snap-Cap Centrifuge Tubes, Plain
Shop alle MBP Molecular BioProducts produkter
Farve:
Neutral
Sterilitet:
Ikke-steril
Steril
Pakningsstørrelse:
Pakke med 1000
Beskrivelse
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Tekniske data
Tekniske data
| Kasket stil | Flat |
| Certificeringer/compliance | Certified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA. |
| Oversigt | Plain, Snap-Cap Centrifuge Tube |
| Kapacitet (metrisk) | 0.6 mL |
| Farve | Neutral |
| Materiale | Polypropylene |
| Emballage | 10 bags of 100 |
| Sterilitet | Steril |
| Uddannet | No |
| Maks. RCF | 25,000 x g |