missing translation for 'onlineSavingsMsg'
Få mere at vide

SN-38 Glucuronide, TRC

Product Code. 30003577
Change view
Click to view available options
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30003577 1 mg
30008822 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003577 Supplier Toronto Research Chemicals Supplier No. TRCS5899801MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121080-63-5
Molekylær formel C28 H28 N2 O11
Molekylvægt (g/mol) 568.53
Synonym β-D-Glucopyranosiduronic acid, (4S)-4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl, β-D-Glucopyranosiduronic acid, 4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl, (S)-, (4S)-4,11-Diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl β-D-glucopyranosiduronic acid, SN 38G, SN-38 glucuronide, 10-Hydroxy-7-ethylcamptothecin 10-β-D-glucuronide, 7-Ethyl-10-hydroxy-20(S)-camptothecin 10-β-D-glucuronide, 7-Ethyl-10-hydroxycamptothecin 10-β-D-glucuronide, (4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione 9-(β-D-glucopyranosiduronide), 11-Ethyl-9-hydroxycamptothecin 9-β-D-glucuronide
SMIL CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O)cc15)[C@@](O)(CC)C(=O)OC4

Specifications

Kemisk navn eller materiale SN-38 Glucuronide
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C28H28N2O11/c1-3-12-13-7-11(40-26-22(33)20(31)21(32)23(41-26)25(35)36)5-6-17(13)29-19-14(12)9-30-18(19)8-16-15(24(30)34)10-39-27(37)28(16,38)4-2/h5-8,20-23,26,31-33,38H,3-4,9-10H2,1-2H3,(H,35,36)/t20-,21-,22+,23-,26+,28-/m0/s1
Formel vægt 568.1693
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.