missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Bio-Techne NBP2-99700AF350
Detalles adicionales : Weight : 0.00970kg
Descripción
SMYD2 Polyclonal specifically detects SMYD2 in Human samples. It is validated for ELISA.Especificaciones
SMYD2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
56950 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 350 | |
EC 2.1.1.43, Histone methyltransferase SMYD2, HSKM-Bzinc finger, MYND domain containing 14, KMT3CEC 2.1.1.-, Lysine N-methyltransferase 3C, MGC119305, SET and MYND domain containing 2, SET and MYND domain-containing protein 2, ZMYND14 | |
Produced in rabbits immunized with purified, recombinant SMYD2 (NP_064582.2; Met 1-His 433). | |
0.1 mL | |
Primary | |
Human | |
Purified |