Learn More
Alfa Aesar™ Smopex™-112v, metal scavenger
Brand:  Alfa Aesar™ 045474.14
Additional Details : Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15402297
  • 2277.31 DKK / 25g
Estimated Shipment date
from Supplier 31-01-2018
Add to basket
Description
Description
  • Recovery of precious metals from catalyst processes, especially Pd and Pt
  • Also, base metals removed: Ag
  • This product is especially suited for aqueous applications
Specifications
Chemical Identifiers
Smopex™-112v metal scavenger
Acrylate based 'alpha'-hydroxyl thiol grafted polyolefin fiber
Specifications
Smopex™-112v metal scavenger
Acrylate based 'alpha'-hydroxyl thiol grafted polyolefin fiber
25g
Powder
Off-white