Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 54.13 cSt (40°C), Paragon Scientific™
Certified in strict accordance with ASTM D2162 at 40 °C and 100 °C under our ISO 17025 and ISO 17034 accreditation and manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids.
Brand:  Paragon Scientific HVS05
Product Code. 16010262
  • 325.00 DKK / 60mL
Description
  • Fully traceable to international standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Certified in strict accordance with ASTM D2162
  • 60mL lower volume cost effective solution
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications
Specifications
60 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 54.13 cSt (40°C), Paragon Scientific™ > Quantity: 60 mL
Nominal values quoted. Certified Values will vary from batch to batch.