Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 23037 cSt (40°C), Paragon Scientific™
Certified in strict accordance with ASTM D2162 at 40 °C and 100 °C under our ISO 17025 and ISO 17034 accreditation and manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids.
Brand:  Paragon Scientific HVS16
Additional Details : Weight : 0.25000kg
Product Code. 16070180
  • 293.00 DKK / 60mL
Estimated Shipment: 24-05-2024
to see stock.
Description
Description
  • Fully traceable to international standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Certified in strict accordance with ASTM D2162
  • 60mL lower volume cost effective solution
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.