missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAGP Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06324AF350
Additional Details : Weight : 0.00970kg
Description
SMAGP Polyclonal specifically detects SMAGP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SMAGP | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
hSMAGP, MGC149453, MGC149454, small cell adhesion glycoprotein, small cell transmembrane and glycosylated protein, small trans-membrane and glycosylated protein, Small transmembrane and glycosylated protein | |
Produced in rabbits immunized with E. coli-derived Human SMAGP fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57228 | |
Store at 4°C in the dark. | |
IgG |