missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad4 Rabbit anti-Human, Mouse, Rat, Clone: 1H7D3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-15673-100UL
Additional Details : Weight : 0.00970kg
Description
Smad4 Monoclonal antibody specifically detects Smad4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP), KnockDownSpecifications
Smad4 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown | |
1H7D3 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:500 - 1:1000, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) 1:500 - 1:1000, Knockout Validated | |
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 320-552 of human Smad4 (NP_005350.1). PEYWCSIAYFEMDVQVGETFKVPSSCPIVTVDGYVDPSGGDRFCLGQLSNVHRTEAIERARLHIGKGVQLECKGEGDVWVRCLSDHAVFVQSYYLDREAGRAPGDAVHKIYPSAYIKVFDLRQCHRQMQQQAATAQAAAAAQAAAVAGNIPGPGSVGGIAPAISLSAAAGIGVDDLRRLCILRMSFVKGWGPDYPRQSIKETPCWIEIHLHRALQLLDEVLHTMPIADPQPLD | |
100 μg | |
Signal Transduction, Stem Cell Signaling Pathway | |
4089 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |