Få mere at vide
Tocris Bioscience™ SM 324405
Potent TLR7 agonist
Brand: Tocris Bioscience™ 5445/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SM 324405 is a potent TLR7 agonist (EC50 = 50 nM). Exhibits selectivity for TLR7 over TLR8. Induces IFN-α and -γ expression in human PBMCs and mouse splenocytes, and inhibits IL-5 production in human PBMCs in vitro. Rapidly metabolized to acid metabolite (t½ = 2.6 min in human plasma).Tekniske data
| Methyl 3-[(6-amino-2-butoxy-7,8-dihydro-8-oxo-9H-purin-9-yl)methyl]benzeneacetate | |
| 677773-91-0 | |
| 50 mg |