missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLK Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-98859JF646
Additional Details : Weight : 0.00970kg
Description
SLK Polyclonal specifically detects SLK in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLK | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
bA16H23.1, CTCL tumor antigen se20-9, EC 2.7.11, EC 2.7.11.1, hSLK, KIAA0204LOSK, Long Ste20-like Kinase, se20-9, Serine/threonine-protein kinase 2, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase (yeast), SNF1 sucrose nonfermenting like kinase, STE20-like kinaseSTE20-like kinase (yeast), STE20-like serine/threonine-protein kinase, STE20-related kinase, Ste20-related serine/threonine kinase, STE20-related serine/threonine-protein kinase, STK2MGC133067 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SLK. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
9748 | |
Store at 4°C in the dark. | |
IgG |