Learn More
Thermo Scientific™ Slideport™ Slide Inserts
Product Code. 17226316 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 pack
5 pack
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17226316 1 pack Each
16644982 5 pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17226316 Supplier Thermo Scientific™ Supplier No. N0101111
 Restricted Item
Forventet forsendelse: 25-11-2026
Image slides on high content screening (HCS) or high content analysis (HCA) readers or on any instrument using a standard microplate.
Image slides on your high content screening (HCS) or high content analysis (HCA) reader or on any instrument in which you use a standard microplate, using Thermo Scientific™ Slideport™ Slideholder.
  • Slideport has been tested to image a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays and beads
  • Image slides as if you were imaging using standard microplates. Any microscope or plate reader that accepts standard microplates will also accept the Slideport
  • Two skirt heights available to accommodate a variety of readers and objectives
  • Automate imaging of your slides by utilizing Slideport's automation compatibility
  • Slideport can be utilized at a variety of temperatures from 4° to 37°C, allowing for flexibility of storage and imaging conditions
  • Compatible with: HCS reader or any instrument that uses a standard microplate, the Thermo Scientific High Content Instruments
  • The Thermo Scientific™ Slideport™ Slide Insert holds microscope slides in a frame that fits Thermo Scientific™ High Content Instruments. Slides can be imaged using many objective lenses due to two available skirt heights.
  • Contents: one Slideport Slide Insert
  • Easily stack slides for automated imaging with or without lids
TRUSTED_SUSTAINABILITY
Specifications
Type Slide Insert
Product Line Slideport
Description Cellomics Slideport Slide Inserts
Compatibility HCS reader or any instrument that uses a standard microplate
For Use With (Equipment) CellInsight CX5 HCS Platform, ArrayScan HCA Reader
Holds 4 slides per insert
Quantity 1 pack