missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SLC6A4/5-HTTLPR/Serotonin transporter Antibody (632705) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB17292JF585
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Flow Cytometry
Tekniske data
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Monoclonal | |
| Janelia Fluor 585 | |
| 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| Human SLC6A4/5-HTTLPR synthetic peptide, Accession # P31645 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 632705 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Neuroscience | |
| 6532 | |
| Store at 4°C in the dark. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion