missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06675
Additional Details : Weight : 0.00970kg
Description
SLC39A14 Polyclonal antibody specifically detects SLC39A14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC39A14 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Immunohistochemistry, Immunohistochemistry-Paraffin 5 to 7.5 μg/mL | |
Irt-like protein 14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, solute carrier family 39 (metal ion transporter), member 14, solute carrier family 39 (zinc transporter), member 14, solute carrier family 39 member 14, zinc transporter ZIP14, ZIP14, ZIP-14, zrt- and Irt-like protein 14 | |
Synthetic 15 amino acid peptide from cytoplasmic domain of human SLC39A14. | |
50 μg | |
Cancer, Endocrinology, Signal Transduction | |
23516 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. |