Learn More
Novus Biologicals™ SLC34A2 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP1-81013PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human SLC34A2. Source: E.coli Amino Acid Sequence: MRSLKPWDAVVSKFTGCFQMRCCCCCRVCCRACCLLCGCPKCCRCSKCCEDLEEAQEGQDVPVKAPETFDNITISREAQGEVPASDSKTECT The SLC34A2 Recombinant Protein Antigen is derived from E. coli. The SLC34A2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP1-81013. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifications
10568 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
SLC34A2 | |
28kDa | |
0.1mL | |
E.Coli | |
MRSLKPWDAVVSKFTGCFQMRCCCCCRVCCRACCLLCGCPKCCRCSKCCEDLEEAQEGQDVPVKAPETFDNITISREAQGEVPASDSKTECT |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unlabeled | |
SLC34A2 | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-81013. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For Research Use Only