missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SLC31A1/CTR1 Antibody (5E6.1B4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80951
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SLC31A1/CTR1 Monoclonal specifically detects SLC31A1/CTR1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Tekniske data
| SLC31A1/CTR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 2 μg/mL, Immunohistochemistry : 5 μg/mL, Immunocytochemistry/Immunofluorescence 0.0763888888888889, Immunohistochemistry-Paraffin : 5 μg/mL | |
| Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 5E6.1B4 | |
| Unconjugated | |
| PBS | |
| SLC31A1 | |
| Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt# O15431] | |
| 0.1 mg | |
| Neuroscience, Neurotransmission, Protein Kinase | |
| 1317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion