Få mere at vide
Novus Biologicals™ SLC22A13 Overexpression Lysate
Artikelnummer. 18079838 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18079838 0.1 mg
1 options
Artikelnummer. 18079838 Leverandør Novus Biologicals™ Leverandørnr. NBP207894
 Begrænset vare
Forventet forsendelse: 09-10-2026
The SLC22A13 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_4247. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens SLC22A13, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only