missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13362
Additional Details : Weight : 0.00970kg
Description
SLC1A7 Polyclonal antibody specifically detects SLC1A7 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC1A7 | |
Polyclonal | |
PBS, 1% BSA, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
AAAT, EAAT5, excitatory amino acid transporter 5 (retinal glutamate transporter), excitatory amino acid transporter 5a long, retinal glutamate transporter, solute carrier family 1 (glutamate transporter), member 7 | |
Recombinant protein encompassing a sequence within the C-terminus region of mouse SLC1A7. The exact sequence is proprietary. | |
100 μg | |
Signal Transduction | |
6512 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |