missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ SLC16A12 Polyclonal Antibody

Artikelnummer. 16380335
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16380335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16380335 Leverandør Invitrogen™ Leverandørnr. PA583379

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: ITLKEDHTTP EQNHVCRTQK EDIKRVSPYS SLTKEWAQTC LCCCLQQEYS Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 62%.

This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SLC16A12
Applikationer Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Koncentration 0.05 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc16a12
Genadgangsnr. Q6ZSM3
Gene Alias 3230401A21; AW210596; CJMG; Creatine transporter 2; CRT2; LOW QUALITY PROTEIN: monocarboxylate transporter 12; MCT 12; MCT12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; RGD1311468; SLC16A12; solute carrier family 16 (monocarboxylic acid transporters), member 12; solute carrier family 16 member 12; solute carrier family 16, member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gensymboler Slc16a12
Værtsarter Rabbit
Immunogen Recombinant protein corresponding to Human SLC16A12. Recombinant protein control fragment (Product #RP-97125).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 387700
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.