missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SLAM/CD150 Antibody (459911) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4330JF585
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Mouse samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Tekniske data
| SLAM/CD150 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
| Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150, Thr25-Pro242, Accession # Q9QUM4 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 459911 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 6504 | |
| Store at 4°C in the dark. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion