Learn More
DWK Life Sciences DURAN™ Long Ground Cone with tip, finished
For ground joints in the high-vacuum range with extension.
Brand: DWK Life Sciences 286166407
Additional Details : Weight : 12.50000kg
Description
The DURAN™ long ground cores with outlet combine the advantages of long ground and outlet cores.
- Ground (finer grinding quality, less roughness depth)
- The larger ground surface is suitable for use in high-vacuum applications.
- With drain
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices | |
22 mm | |
NS 24/40 | |
1.8 mm |