Få mere at vide
Tocris Bioscience™ SJF 8240
Beskrivelse
SJF 8240 is a c-MET PROTAC™Degrader. Comprises MET inhibitor foretinib (GSK 1363089; Cat. No. 6056) joined by a linker to a von Hippel-Lindau (VHL) recruiting ligand. Degrades c-MET within 6 hours in vitro. Inhibits agonist-driven AKT phosphorylation and GTL16 cell proliferation (IC50 = 66.7 nM). Also degrades exon-14-deleted c-MET in Hs746T cells. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(3-Fluoro-4-((7-(3-(3-(3-((-1-((2 S,4 R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)propoxy)propoxy)-6-methoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dic |
| CAS | 2230821-68-6 |
| Mængde | 5 mg |
| Mål | Active Degraders |
| Molekylær formel | C58H65F2N7O11S |
| Renhed | 0.98 |