missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ SJ 10542
GREENER_CHOICE

Artikelnummer. 30000433
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30000433 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30000433 Leverandør Tocris Bioscience™ Leverandørnr. 7727/2

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Potent and selective JAK2/3 Degrader (PROTAC™)

SJ 10542 is a potent and selective JAK2/3 Degrader (PROTAC™) (DC50 values are 11 nM and 14 nM for JAK3 and JAK2, respectively), it uses a phenyl glutarimide (PG) ligand as the cereblon (CRBN) recruiter. SJ 10542 displays high selectivity over GSPT1 and other members of the JAK family. SJ 10542 is active in cytotoxicity assays with PDX models (IC50 values <120 nM).

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-((4-(1-(3-(Cyanomethyl)-1-(ethylsulfonyl)azetidin-3-yl)-1H-pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidin-2-yl)amino)-N-(3-(4-(4-(2,6-dioxopiperidin-3-yl)phenyl)piperazin-1-yl)propyl)benzamide
CAS 2789678-92-6
Mængde 2 mg
Mål Active Degraders
Molekylær formel C41H46N12O5S
Renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.