Learn More
Duran™ Sintered Glass Filter Disc, 20mm Diameter
Duran™ glass filter for filtration both during analytical as well as preparative work.
Brand:  Duran™ 251520004
367.65 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.02000kg
Product Code. 15252019
  • 387.00 DKK / Pack of 10
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Rim unfused and centered
  • Surface unprocessed
  • Available in packs of ten
Specifications
Specifications
Sintered Glass
160 to 250 μm
10Pack