Learn More
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe
Use these syringes for veterinary medicine applications. Poulten Graf™ Single Use Luer-Tip Two-Part Syringe are PVC-free and non-toxic, sterile packed and depatched in blister packs. The single-use syringes are extremely tight, and have a transparent, smooth operating cylinder.
398.28 DKK - 977.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | CE |
Material | Polypropylene |
Description | Screen printed graduation |
Product Type | Luer-tip 2-part Syringe |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10730353
|
Poulten Graf™
7 41008 |
20 to 24 mL |
977.00 DKK
Pack of 100 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10269521
|
Poulten Graf™
7 41004 |
5 to 6 mL |
483.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10076041
|
Poulten Graf™
7 41006 |
10 to 12 mL |
667.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10640513
|
Poulten Graf™
7 41012 |
50 to 60 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11329553
|
Poulten Graf™
7 41012 |
50 to 60 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10279521
|
Poulten Graf™
7 41001 |
2 to 3 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Produced in accordance with DIN 13098
- Available in a variety of sizes, with and without needle
Specifications
Autoclavable | |
Polypropylene | |
Luer-tip 2-part Syringe | |
Luer Tip | |
Sterile |
CE | |
Screen printed graduation | |
Yes | |
Yes |