missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Simvastatin Hydroxy Acid Ammonium Salt, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Simvastatin Hydroxy Acid, Ammonium Salt |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C25 H39 O6 . H4 N |
| Mængde | 100 mg |
| Synonym | 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, ammonium salt (1:1), (βR,δR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monoammonium salt, (βR,δR,1S,2S,6R,8S,8aR)- (9CI), 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monoammonium salt, [1S-[1α(βS*,δS*),2α,6β,8β,8aα]]-, Ammonium simvastatin, Simvastatin acid ammonium salt, Simvastatin ammonium salt, Simvastatin carboxylic acid ammonium salt, (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic acid ammonium salt, Tenivastatin ammonium salt |
| InChI formel | InChI=1S/C25H40O6.H3N/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);1H3/t15-,16-,18+,19+,20-,21-,23-;/m0./s1 |
| SMIL | [NH4+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| Formel vægt | 453.309 |
| Procent renhed | >95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?