Få mere at vide
Simurosertib, MedChemExpress
Artikelnummer. 16495937
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16495937 5 mg
16483208 10 mg
16479756 50 mg
16440046 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16495937 Leverandør Medchem Express Leverandørnr. HY100888
MedChemExpress Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM. Simurosertib has anti-cancer activity.
Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM. Simurosertib has anti-cancer activity.
Tekniske data
Kemisk navn eller materiale Simurosertib
Fysisk form Solid
CAS 1330782-76-7
Farve Light Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C17H19N5OS
Til brug med (applikation) Cancer-Kinase/protease
Synonym TAK-931
Opløselighedsinformation DMSO : 75 mg/mL (219.66 mM; Need ultrasonic)
SMIL O=C1C2=C(C=C(C3=CNN=C3C)S2)NC([C@H]4N(CC5)CCC5C4)=N1
Molekylvægt (g/mol) 341.43
Formel vægt 341.43
Procent renhed 98.1%