Beskrivelse
User-friendly to both patients and laboratory personnel
- Innovative cap features a large opening for easy collection
- A leakproof-cap liner ensures spill-free sample transport
- Vertical and horizontal graduations allow for accurate and easy reading, eliminating transfer to graduated cylinder for volume measurement
- Containers can be subjected to freezing, thawing and EtO gas sterilization without causing changes in materials or physical appearance
- Resistant to hydrochloric acid
- Convenient refrigerator storage with less wasted space
- Containers are available in 3L and 4L tilt size
- The 4L bottle allows a clean and easy transfer to tubes by simply tilting the container forward without lifting
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Produkttitel
Simport™ Scientific Urine Collection URISAFE™ Containers