Learn More
Simport™ Scientific PCRack™
The PCRack™ is designed especially for storing and working with PCR samples.
POA
Specifications
Product Type | Microtube Rack |
---|---|
Autoclavable | Not Autoclavable |
Holds | Tubes (0.2 mL) |
Material | Polypropylene |
Positions | 96 |
Description
- Base and cover constructed of autoclavable polypropylene
- Temperature range from -90°C to +121°C
- Accommodates 0.2 mL tubes, along with strips of 8 or 12 tubes and 96-well PCR plates
- Alphanumeric configuration, indexed from A to H vertically and 1 to 12 horizontally
- Cover has easy to read numbers and letters for secure identification of contents
- Can be horizontally attached to each other, or stacked on top of another
- Available in 6 colors
Specifications
Microtube Rack | |
Tubes (0.2 mL) | |
96 | |
-90°C to +121°C |
Not Autoclavable | |
Polypropylene | |
20/Cs. |