Få mere at vide
Simport™ Scientific E-Z LOAD™ Histosette™ II Tissue Cassette Lids
These cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
Beskrivelse
Beskrivelse
  • For use with Micro Writer™, Lamb™, and Shur/Mark™ cassette labeling instruments
  • Stacks of 50 cassettes each; case contains two boxes of 20 stacks
  • Bases and covers must be ordered separately
Videoer
SDS
Dokumenter
Dokumenter
Produktcertifikater