Learn More
Motic™ Simple Darkfield Slider (10X - 40X)
Enhances contrast. Motic™ Simple Darkfield Slider (10X - 40X) is compatible with all BA microscopes
Brand: Motic™ 1101001900542
Additional Details : Weight : 0.80000kg
Description
- Enhances contrast
- 10X - 40X
- For use with any BA Series microscope
Order Info
dimForUseWithEquipment
Specifications
Simple Darkfield Slider | |
BA210 and BA310 Series Upright Compound Microscope | |
1 Ea. |