Få mere at vide
Simendan, TRC
Artikelnummer. 30134039
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134039 5 g
17974275 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134039 Leverandør Toronto Research Chemicals Leverandørnr. TRCS4660005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131741-08-7
Molekylær formel C14H12N6O
Molekylvægt (g/mol) 280.28
Synonym [[4-(1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]hydrazono]-propanedinitrile
IUPAC navn 2-[[4-(4-methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl)phenyl]hydrazinylidene]propanedinitrile
SMIL O=C1CC(C(C2=CC=C(N/N=C(C#N)/C#N)C=C2)=NN1)C
Tekniske data
Kemisk navn eller materiale Simendan
Renhedsgrad noter HPLC
Mængde 5 g
InChI formel InChI=1S/C14H12N6O/c1-9-6-13(21)19-20-14(9)10-2-4-11(5-3-10)17-18-12(7-15)8-16/h2-5,9,17H,6H2,1H3,(H,19,21)
Formel vægt 280.1073
Procent renhed >95%