missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Simazine D10 100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | Simazine D10 |
| CAS | 220621-39-6 |
| Opløsningsmiddel eller Matrix | Acetone |
| Molekylær formel | C7 2H10 H2 Cl N5 |
| Mængde | 1 mL |
| Synonym | Simazine D10 |
| InChI formel | InChI=1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)/i1D3,2D3,3D2,4D2 |
| SMIL | [2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC([2H])([2H])C([2H])([2H])[2H])n1 |
| IUPAC navn | 6-chloro-2-N,4-N-bis(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine |
| Molekylvægt (g/mol) | 211.72 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?