Få mere at vide
Silychristin, TRC
Artikelnummer. 17991145
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991145 5 mg
17910845 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991145 Leverandør Toronto Research Chemicals Leverandørnr. TRCS4658805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33889-69-9
Molekylær formel C25 H22 O10
Molekylvægt (g/mol) 482.44
Synonym 4H-1-Benzopyran-4-one, 2-[(2R,3S)-2,3-dihydro-7-hydroxy-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-5-benzofuranyl]-2,3-dihydro-3,5,7-trihydroxy-, (2R,3R)-, 4-Chromanone, 2-[2,3-dihydro-7-hydroxy-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-5-benzofuranyl]-3,5,7-trihydroxy-, (2S,3S)-(+)- (8CI), 4H-1-Benzopyran-4-one, 2-[2,3-dihydro-7-hydroxy-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-5-benzofuranyl]-2,3-dihydro-3,5,7-trihydroxy-, [2R-[2α(2R*,3S*),3β]]-, (2R,3R)-2-[(2R,3S)-2,3-Dihydro-7-hydroxy-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-5-benzofuranyl]-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, (+)-Silychristin, Silichristin, Silicristin, Silychristin A, Silycristin, Silymarin II
IUPAC navn (2R,3R)-3,5,7-trihydroxy-2-[(2R,3S)-7-hydroxy-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydro-1-benzofuran-5-yl]-2,3-dihydrochromen-4-one
SMIL COc1cc(ccc1O)[C@@H]2Oc3c(O)cc(cc3[C@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O
Tekniske data
Kemisk navn eller materiale Silychristin
Mængde 5 mg
InChI formel InChI=1S/C25H22O10/c1-33-18-6-10(2-3-15(18)28)23-14(9-26)13-4-11(5-17(30)25(13)35-23)24-22(32)21(31)20-16(29)7-12(27)8-19(20)34-24/h2-8,14,22-24,26-30,32H,9H2,1H3/t14-,22+,23+,24-/m1/s1
Formel vægt 482.1213