Få mere at vide
Silvestrol, MedChemExpress
Beskrivelse
Silvestrol is a eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from the fruits and twigs of Aglaia foveolata. Silvestrol induces autophagy and caspase-mediated apoptosis.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Silvestrol |
| CAS | 697235-38-4 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C34H38O13 |
| Mængde | 2 mg |
| Synonym | (-)-Silvestrol |
| Opløselighedsinformation | DMSO : ≥ 6.6 mg/mL (10.08 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMIL | O=C([C@H]([C@H]1C2=CC=CC=C2)[C@@H](O)[C@]3(O)[C@@]1(C4=CC=C(OC)C=C4)OC5=CC(O[C@@H]6O[C@@H]([C@H](O)CO)CO[C@H]6OC)=CC(OC)=C35)OC |
| Molekylvægt (g/mol) | 654.66 |
| Vis mere |