Learn More
Silver trifluoromethanesulfonate, 99+%, Thermo Scientific Chemicals
Catalog Number p-4510609
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
2923-28-6
CAgF3O3S
256.93
MFCD00013226
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
76223
silver;trifluoromethanesulfonate
[Ag+].[O-]S(=O)(=O)C(F)(F)F
2923-28-6
CAgF3O3S
256.93
MFCD00013226
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
76223
silver;trifluoromethanesulfonate
[Ag+].[O-]S(=O)(=O)C(F)(F)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 356.0°C
Color White to Beige
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CF3SO3Ag
Quantity 10 g
Merck Index 09,302
Solubility Information Solubility in water: soluble.
Formula Weight 256.92
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Silver trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Silver trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.