missing translation for 'onlineSavingsMsg'
Learn More
Silver Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ Product Code.: 10152492

Silver Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Product Code. p-7073221 Shop All Fisher Chemical Products
100 mL, HDPE plastic bottle, 100mL
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber glass bottle
HDPE plastic bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10152492

Brand: Fisher Chemical J/8064/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Packaging HDPE plastic bottle
Physical Form Liquid
Color Colorless
pH 1
IUPAC Name silver(1+) nitrate
Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • Silver (I) single element solution 1,000
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P273-Avoid release to the environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.