Learn More
Linden™ Silver Oxide Batteries
Description
- Supplied in small button form
- High energy to weight ratio
Specifications
Specifications
| Product Type | Silver Oxide Button Battery |
| Cell Type | Button |
| Quantity | 4 Pk. |
| Voltage | 1.55 V |
| Amperage | 130 mAh capacity |