missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, ReagentPlus™, Honeywell

Catalog Number p-7072315
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
Quantity 5 g
Linear Formula AgNO3
UN Number UN1493
Formula Weight 169.87g/mol
Percent Purity ≥99% (titration)
Grade ReagentPlus™
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'