missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Iodide, 99%, Honeywell™ Fluka™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Yellow |
Physical Form | Solid |
Quantity | 100 g |
UN Number | UN3077 |
Formula Weight | 234.77g/mol |
Percent Purity | 99% |
Packaging | Glass Bottle |
Density | 5.68g/mL at 25°C |
Chemical Name or Material | Silver Iodide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'