missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver cyanate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 98% |
Beilstein | 3655040 |
Solubility Information | Soluble in ammonia,nitric acid,potassium cyanide,ammonium hydroxide. Insoluble in alcohol and dilute acids. |
Formula Weight | 149.89 |
Odor | Odorless |
Sensitivity | Light sensitive |
Density | 4 g/cm3 at 20°C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'