Learn More
uvex Silv-Air 5200 Premium FFP2 Flat-fold Mask
Product Code. 31910425 Shop All Uvex Products
Change view
Click to view available options
Size:
Medium
Unit Size:
Each
Product Code. Size Unit Size
31910425 Medium Each
1 options
Product Code. 31910425 Supplier uvex Supplier No. 8735200
  • ONLINE EXCLUSIVE PRICE
    595.00 DKK
    556.84 DKK / Pakke med 1
    Spare 38.16 DKK
    6% Slukket
Forventet forsendelse: 27-11-2026
The Silv-Air premium product line comprises classic, white folding respirators that adapt to different face shapes.
  • The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
  • Particle-filtering folding mask
  • Individually packed
  • White color
  • Also available with exhalation valve and carbon filter
  • Flexible and adjustable nose clip
  • Soft sealing lip guarantees a secure, comfortable sealed position of the mask
  • Soft material edges for high wearing comfort
  • Adjustable head band offering easy handling and comfortable fit
  • Certified FFP2 face mask to EN 149:2001+A1:2009 D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
  • Respirators are not suitable for people with beards, sideburns or deep scars near the seals
TRUSTED_SUSTAINABILITY
Specifications
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polyester
Product Type Particulate Respirator
Size Medium
With Exhalation Valve No