Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand: Deutsch Neumann™ 1010550
Additional Details : Weight : 0.09900kg
Description
- Acc. to DIN 12 871
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
Transparent | |
50.5 mm | |
1/Ea. |
Stopper | |
59.5 mm | |
Silicone |