Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand:  Deutsch Neumann™ 1010594
Additional Details : Weight : 0.56000kg
 View more versions of this product
Product Code. 10406321
  • 580.00 DKK / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
Transparent
94 mm
1/Ea.
Stopper
107 mm
Silicone