Learn More
Macherey-Nagel™ Silicone Standard Septa
Brand:  Macherey-Nagel™ 702613
 View more versions of this product
Product Code. 10361102
  • 405.00 DKK / Pack of 50
Only null left
Estimated Shipment: 30-06-2025
Specifications
Standard Septum
60° Shore A
50 Pack
13 mm
Silicone
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Silicone Standard Septa > O.D.: 13mm