missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ BPX70 GC Capillary Column

SGE BPX70 GC Capillary Column is an MS premium, low bleed column with high polarity specifically designed for fatty acid methyl ester (FAME) analysis

Brand:  Trajan Scientific and Medical™ 054622

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12624955

  • 5480.00 DKK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

  • High temperature
  • Custom designed for separation of FAMEs
  • Ideal for cis/trans isomer separation
  • Polar phase
Specifications

Specifications

0.25 mm
BPX70
50°C to 250/260°C
30 mm
GC Capillary Column
250/260°C
0.25 mm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions