Learn More
Trajan Scientific and Medical™ GP GC Septa
Combines significantly longer injection life, low bleed and low injection port adhesion
Brand:  Trajan Scientific and Medical™ 041824
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10441391
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Designed for non-demanding, routine applications
  • Reliable seal against carrier gas pressure in inlet
  • No bleed or contamination into GC system
  • Maximum operating temperature of 275°C
Specifications
Specifications
9 mm
275°C
GP Septum
Silicone
Yes
50 Pack