Learn More
Chem Lab™ Silicon Oil
Product Code. 13435819
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
Product Code. Volume (Metric) Unit Size
13435819 1 L 1 L
1 options
Product Code. 13435819 Supplier Chem Lab™ Supplier No. CL00.1918.1000
  • ONLINE EXCLUSIVE PRICE
    767.00 DKK
    537.32 DKK / 1 L
    Spare 229.68 DKK
    29% Slukket
View all promotions
 Restricted Item
Forventet forsendelse: 05-01-2027