Learn More
Silicagel Pearls with Orange Indicator, Chem-Lab
Product Code. 11953315
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
Product Code. Quantity Unit Size
11953315 1 Pk. Each
1 options
Product Code. 11953315 Supplier Chem-Lab Supplier No. CL00.1920.1000
  • ONLINE EXCLUSIVE PRICE
    562.00 DKK
    372.39 DKK / Pakke med 1
    Spare 189.61 DKK
    33% Slukket
 Restricted Item
Forventet forsendelse: 14-12-2026
Changes color as desiccant adsorbs moisture. Silicagel Pearls with Orange Indicator, Chem-Lab are silicon dioxide beads that readily adsorb water and protect processes and products from the effects of humidity and moisture. Pearls are 2.5–5mm in size and are orange when in an anhydrous state.
Silica gel's high specific surface area allows it to adsorb water readily, making it useful as a desiccant (drying agent). In chemistry, silica gel is used in chromatography as a stationary phase. This product contains an orange indicator that changes color as maximum adsorbance is achieved, so that the gel can be replaced.
Specifications
Physical Form Solid
Quantity 1 Pk.