missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator, Thermo Scientific Chemicals

Catalog Number p-7058567
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O
This item is not returnable. View return policy
63231-67-4
H2O3Si
78.098
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O

TLC plate preparation

Silica gel is primarily used in plate preparation of thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form 12 Micron APS,S.A. 500-600m
pH 6.5 to 7.5
Material Silica Gel
Quantity 50 g
Particle Size 500 to 600m2/g, 60Å
Solubility Information Insoluble in water.
Grade TLC High Purity
Surface Area 500 to 600m2/g
Chemical Name or Material Silica gel, With gypsum binder and fluorescent indicator

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'