Få mere at vide
Sildenafil-d3, TRC
Artikelnummer. 30409096
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30409096 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409096 Leverandør Toronto Research Chemicals Leverandørnr. TRCS4350025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sildenafil-d3
CAS 1126745-90-1
Molekylær formel C22 D3 H27 N6 O4 S
Mængde 5 mg
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one','5-[2-ethoxy-5-[[4-(methyl-d3)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Sildenafil-d3, 5-[2-Ethoxy-5-[[4-(methyl-d3)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Sildenafil-D3 (N-methyl-D3-piperazine)
InChI formel InChI=1S/C22H30N6O4S/c1-5-7-17-19-20(27(4)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29)/i3D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)S(=O)(=O)c2ccc(OCC)c(c2)C3=Nc4c(CCC)nn(C)c4C(=O)N3
IUPAC navn 5-[2-ethoxy-5-[4-(trideuteriomethyl)piperazin-1-yl]sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molekylvægt (g/mol) 477.595
Formel vægt 477.224